N-Methylbenzolsulfonamid

Lieferant: EHRENSTORFER

C15083780
EHERC15083780EA 56 EUR
EHERC15083780
N-Methylbenzolsulfonamid
N-Methylbenzolsulfonamid
Formel: C₇H₉NO₂S
Molecular Weight: 171,21 g/mol
MDL: MFCD00025023
CAS-Nummer: 5183-78-8
EINECS: 225-965-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR