2-Methylbenzofuran
Lieferant: EHRENSTORFER
C15083785
EHERC15083785EA
69
EUR
EHERC15083785
2-Methylbenzofuran
2-Methylbenzofuran
Formel:
C₉H₈O Molecular Weight: 132,16 g/mol |
MDL:
MFCD00005850 CAS-Nummer: 4265-25-2 EINECS: 224-249-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...