5-Methylbenzotriazol
Lieferant: EHRENSTORFER
Synonyme:
5-Methyl-1H-benzotriazole
C15083795
EHERC15083795EA
64
EUR
EHERC15083795
5-Methylbenzotriazol
5-Methylbenzotriazol
Formel:
C₇H₇N₃ Molecular Weight: 133,15 g/mol Siedepunkt: 361 °C (1013 hPa) Schmelzpunkt: 85 °C |
MDL:
MFCD00005702 CAS-Nummer: 136-85-6 EINECS: 205-265-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...