4,4'-Methylenbis(2-chloranilin)
Lieferant: EHRENSTORFER
C15087500
EHERC15087500EA
51
EUR
EHERC15087500
4,4'-Methylenbis(2-chloranilin)
4,4'-Methylenbis(2-chloranilin)
Formel:
C₁₃H₁₂Cl₂N₂ |
MDL:
MFCD00047829 CAS-Nummer: 101-14-4 EINECS: 202-918-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...