2-Methyl-4(5)-nitroimidazol

Lieferant: EHRENSTORFER

Synonyme: 2-Methyl-4-nitroimidazol, 2-Methyl-4-nitro-1H-imidazol

C15107000
EHERC15107000EA 64 EUR
EHERC15107000
2-Methyl-4(5)-nitroimidazol
2-Methyl-4(5)-nitroimidazol
Formel: C₄H₅N₃O
Molecular Weight: 127,10 g/mol
MDL: MFCD00005191
CAS-Nummer: 696-23-1
EINECS: 211-790-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR