3-Nitro-o-kresol

Lieferant: EHRENSTORFER

Synonyme: 1-Hydroxy-2-methyl-3-nitrobenzol, 2-Methyl-3-nitrophenol, 2-Hydroxy-6-nitrotoluol

C15108500
EHERC15108500EA 38 EUR
EHERC15108500
3-Nitro-o-kresol
3-Nitro-o-kresol
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 149…152 °C
MDL: MFCD00007241
CAS-Nummer: 5460-31-1
EINECS: 226-739-0
UN: 2446
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR