4-(Methylsulfonyl)-2-nitrobenzoesäure
Lieferant: EHRENSTORFER
C15143800
EHERC15143800EA
135
EUR
EHERC15143800
4-(Methylsulfonyl)-2-nitrobenzoesäure
4-(Methylsulphonyl)-2-nitrobenzoic acid
Formel:
C₈H₇NO₆S |
MDL:
MFCD01941299 CAS-Nummer: 110964-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...