2-(Methylthio)benzothiazol

Lieferant: EHRENSTORFER

C15144200
EHERC15144200EA 103 EUR
EHERC15144200
2-(Methylthio)benzothiazol
2-(Methylthio)benzothiazole
Formel: C₈H₇NS₂
MDL: MFCD00005784
CAS-Nummer: 615-22-5
EINECS: 210-417-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR