6-Methyl-2-thiouracil
Lieferant: EHRENSTORFER
Synonyme:
4-Hydroxy-2-mercapto-6-methylpyrimidin, Methylthiouracil
C15144300
EHERC15144300EA
44
EUR
EHERC15144300
6-Methyl-2-thiouracil
6-Methyl-2-thiouracil
Formel:
C₅H₆N₂OS Molecular Weight: 142.18 g/mol Siedepunkt: 351 Dichte: 1.519 g/cm³ (20 °C) |
MDL:
MFCD00006040 CAS-Nummer: 56-04-2 EINECS: 200-252-3 Merck Index: 12,06210 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...