Metoxuron (3-(3-Chlor-4-methoxyphenyl)-1,1-dimethylharnstoff)

Lieferant: EHRENSTORFER

C15180000
EHERC15180000EA 42 EUR
EHERC15180000
Metoxuron (3-(3-Chlor-4-methoxyphenyl)-1,1-dimethylharnstoff)
Metoxuron (3-(3-Chlor-4-methoxyphenyl)-1,1-dimethylharnstoff)
Formel: C₁₀H₁₃ClN₂O₂
CAS-Nummer: 19937-59-8
EINECS: 243-433-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR