Monocrotophos (Dimethyl-1-methyl-2-(methylcarbamoyl)vinylphosphat)
Lieferant: EHRENSTORFER
C15300000
EHERC15300000EA
46
EUR
EHERC15300000
Monocrotophos (Dimethyl-1-methyl-2-(methylcarbamoyl)vinylphosphat)
Monocrotophos (Dimethyl-1-methyl-2-(methylcarbamoyl)vinylphosphat)
Formel:
C₇H₁₄NO₅P Molecular Weight: 223,17 g/mol |
CAS-Nummer:
6923-22-4 EINECS: 230-042-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...