Monolinuron (N-(4-Chlorphenyl)-N'-methoxy,N'-methylharnstoff)

Lieferant: EHRENSTORFER

Synonyme: 3-(4-Chlorophenyl)-1-methoxy-1-methylurea, Monolinuron

C15310000
EHERC15310000EA 61 EUR
EHERC15310000
Monolinuron (N-(4-Chlorphenyl)-N'-methoxy,N'-methylharnstoff)
Monolinuron (N-(4-Chlorphenyl)-N'-methoxy,N'-methylharnstoff)
Formel: C₉H₁₁ClN₂O₂
MDL: MFCD00055264
CAS-Nummer: 1746-81-2
EINECS: 217-129-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR