Monuron (N-(4-Chlorphenyl)-N',N'-dimethylharnstoff)

Lieferant: EHRENSTORFER

C15320000
EHERC15320000EA 47 EUR
EHERC15320000
Monuron (N-(4-Chlorphenyl)-N',N'-dimethylharnstoff)
Monuron (N-(4-Chlorphenyl)-N',N'-dimethylharnstoff)
Formel: C₉H₁₁ClN₂O
Molecular Weight: 198,65 g/mol
MDL: MFCD00018556
CAS-Nummer: 150-68-5
EINECS: 205-766-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR