Neomycinsulfat

Lieferant: EHRENSTORFER

Synonyme: Neomycin Sulfatsalz

C15500900
EHERC15500900EA 60 EUR
EHERC15500900
Neomycinsulfat
Neomycinsulfat
Formel: C₂₃H₄₆N₆O₁₃·3H₂SO₄
Molecular Weight: 908,87 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150827
CAS-Nummer: 1405-10-3
EINECS: 215-773-1
Merck Index: 12,06542

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR