6-Hydroxynicotinsäure
Lieferant: EHRENSTORFER
Synonyme:
6-Hydroxypyridin-3-carbonsäure, 6-Hydroxy-3-pyridincarbonsäure
C15521080
EHERC15521080EA
69
EUR
EHERC15521080
6-Hydroxynicotinsäure
6-Hydroxynicotinsäure
Formel:
C₆H₅NO₃ Schmelzpunkt: 304…306 °C |
MDL:
MFCD00006277 CAS-Nummer: 5006-66-6 EINECS: 225-682-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...