2-Nitrodiphenylamin

Lieferant: EHRENSTORFER

Synonyme: 2-Nitro-N-phenylanilin

C15559200
EHERC15559200EA 41 EUR
EHERC15559200
2-Nitrodiphenylamin
2-Nitrodiphenylamin
Formel: C₁₂H₁₀N₂O₂
Molecular Weight: 214,22 g/mol
CAS-Nummer: 119-75-5
EINECS: 204-348-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR