Nitrofen
Lieferant: EHRENSTORFER
Synonyme:
2,4-Dichloro-1-(4-nitrophenoxy)benzene
C15560000
EHERC15560000EA
55
EUR
EHERC15560000
Nitrofen
Nitrofen
Formel:
C₁₂H₇Cl₂NO₃ Molecular Weight: 284.1 g/mol |
MDL:
MFCD00128026 CAS-Nummer: 1836-75-5 EINECS: 217-406-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...