4-Nitroimidazol
Lieferant: EHRENSTORFER
|
Achtung
|
Synonyme:
4-Nitro-1H-imidazol
C15588300
EHERC15588300EA
58
EUR
EHERC15588300
4-Nitroimidazol
4-Nitroimidazol
Formel:
C₃H₃N₃O₂ Molecular Weight: 113,08 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005196 CAS-Nummer: 3034-38-6 EINECS: 221-224-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...