4-Nitrophenol-[2,3,5,6-D4]
Lieferant: EHRENSTORFER
|
Achtung
|
Synonyme:
4-Nitrophenol-[2,3,5,6-²H₄]
C15590404
EHERC15590404EA
200
EUR
EHERC15590404
4-Nitrophenol-[2,3,5,6-D4]
4-Nitrophenol-[2,3,5,6-D4]
Formel:
C₆HD₄NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143352 CAS-Nummer: 93951-79-2 UN: 1663 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...