Nonadecansäuremethylester

Lieferant: EHRENSTORFER

Synonyme: Nonadecanoic acid methyll ester, Methyl Nonadecanoate

C15622360
EHERC15622360EA 39 EUR
EHERC15622360
Nonadecansäuremethylester
Nonadecanoic acid methyl ester
Formel: C₂₀H₄₀O₂
Molecular Weight: 312,54 g/mol
MDL: MFCD00009011
CAS-Nummer: 1731-94-8
EINECS: 217-056-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR