Nystatin

Lieferant: EHRENSTORFER
Achtung

Synonyme: Fungicidin, Mycostatin

C15661000
EHERC15661000EA 49 EUR
EHERC15661000
Nystatin
Nystatin
Formel: C₄₇H₇₅NO₁₇
Molecular Weight: 926,09 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD25541651
CAS-Nummer: 1400-61-9
EINECS: 215-749-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR