2-Octinsäuremethylester
Lieferant: EHRENSTORFER
Synonyme:
Methyl-2-octinoat
C15715700
EHERC15715700EA
42
EUR
EHERC15715700
2-Octinsäuremethylester
2-Octynoic acid methyl ester
Formel:
C₉H₁₄O₂ Molecular Weight: 154,21 g/mol |
MDL:
MFCD00009530 CAS-Nummer: 111-12-6 EINECS: 203-836-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...