Oxytetracyclin Hydrochlorid
Lieferant: EHRENSTORFER
C15820000
EHERC15820000EA
51
EUR
EHERC15820000
Oxytetracyclin Hydrochlorid
Oxytetracyclin Hydrochlorid
Formel:
C₂₂H₂₅ClN₂O₉ Molecular Weight: 496,90 g/mol |
MDL:
MFCD00135815 CAS-Nummer: 2058-46-0 EINECS: 218-161-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...