2-Ethylhexyl-4-(dimethylamino)benzoat
Lieferant: EHRENSTORFER
Synonyme:
2-Ethylhexyl 4-(dimethylamino)benzoate
C15841000
EHERC15841000EA
99
EUR
EHERC15841000
2-Ethylhexyl-4-(dimethylamino)benzoat
2-Ethylhexyl-4-(dimethylamino)benzoat
Formel:
C₁₇H₂₇NO₂ Molecular Weight: 277,40 g/mol |
MDL:
MFCD00017526 CAS-Nummer: 21245-02-3 EINECS: 244-289-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...