4-Bromdiphenylether (BDE Nr. 3)
Lieferant: EHRENSTORFER
Synonyme:
4-Bromodiphenyl ether, 1-Bromo-4-phenoxybenzene
C15898003
EHERC15898003EA
44
EUR
EHERC15898003
4-Bromdiphenylether (BDE Nr. 3)
4-Bromdiphenylether (BDE Nr. 3)
Formel:
C₁₂H₉BrO Molecular Weight: 249,11 g/mol |
MDL:
MFCD00000094 CAS-Nummer: 101-55-3 EINECS: 202-952-4 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...