Decabromdiphenylether (BDE Nr. 209)

Lieferant: EHRENSTORFER

Synonyme: Bis(pentabromophenyl) ether

C15898209
EHERC15898209EA 43 EUR
EHERC15898209
Decabromdiphenylether (BDE Nr. 209)
Decabromdiphenylether (BDE Nr. 209)
Formel: C₁₂Br₁₀O
MDL: MFCD00000059
CAS-Nummer: 1163-19-5
EINECS: 214-604-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR