Heptadecafluoroctansulfonamid

Lieferant: EHRENSTORFER

C15987110
EHERC15987110EA 154 EUR
EHERC15987110
Heptadecafluoroctansulfonamid
Heptadecafluoroctansulfonamid
Formel: C₈H₂F₁₇NO₂S
MDL: MFCD03094345
CAS-Nummer: 754-91-6
EINECS: 212-046-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR