Kaliumheptadecafluoroctan-1-sulfonat
Lieferant: EHRENSTORFER
C15987122
EHERC15987122EA
92
EUR
EHERC15987122
Kaliumheptadecafluoroctan-1-sulfonat
Kaliumheptadecafluoroctan-1-sulfonat
Formel:
C₈F₁₇KO₃S |
MDL:
MFCD00066407 CAS-Nummer: 2795-39-3 EINECS: 220-527-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...