DL-α-Phenylalanin
Lieferant: EHRENSTORFER
Synonyme:
(±)-2-Amino-3-phenylpropansäure, H-DL-Phe-OH, 3-Phenyl-DL-alanin, DL-Phenylalanin, (±)-2-Amino-3-phenylpropionic acid, (±)-2-Amino-3-phenylpropionsäure
C16055000
EHERC16055000EA
50
EUR
EHERC16055000
DL-α-Phenylalanin
DL-α-Phenylalanin
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol Siedepunkt: 313 °C (1013 hPa) Schmelzpunkt: 173 °C |
MDL:
MFCD00064225 CAS-Nummer: 150-30-1 EINECS: 205-756-7 Merck Index: 13,07355 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...