Benzylbutylphthalat-[Ring-D4]

Lieferant: EHRENSTORFER

Synonyme: Benzyl butyl phthalate-[ring-²H₄]

C16168010
EHERC16168010EA 106 EUR
EHERC16168010
Benzylbutylphthalat-[Ring-D4]
Benzylbutylphthalat-[Ring-D4]
Formel: C₁₉H₁₆D₄O₄
CAS-Nummer: 93951-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR