Dibutylphthalat-[Ring-D4]
Lieferant: EHRENSTORFER
|
Gefahr
|
Synonyme:
Dibutyl phthalate-[ring-²H₄]
C16171010
EHERC16171010EA
123
EUR
EHERC16171010
Dibutylphthalat-[Ring-D4]
Dibutylphthalat-[Ring-D4]
Formel:
C₁₆H₁₈D₄O₄ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
93952-11-5 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...