Bis(2-ethylhexyl)phthalat-[Ring-D4]
Lieferant: EHRENSTORFER
|
Gefahr
|
Synonyme:
Bis(2-ethylhexyl) phthalate-[ring-²H₄]
C16173010
EHERC16173010EA
204
EUR
EHERC16173010
Bis(2-ethylhexyl)phthalat-[Ring-D4]
Bis(2-ethylhexyl)phthalat-[Ring-D4]
Formel:
C₂₄H₃₄D₄O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143759 CAS-Nummer: 93951-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...