Bis(2-methoxyethyl)phthalat

Lieferant: EHRENSTORFER

Synonyme: Bis(2-methoxyethyl) phthalate

C16174400
EHERC16174400EA 42 EUR
EHERC16174400
Bis(2-methoxyethyl)phthalat
Bis(2-methoxyethyl)phthalat
Formel: C₁₄H₁₈O₆
Molecular Weight: 282,29 g/mol
MDL: MFCD00042842
CAS-Nummer: 117-82-8
EINECS: 204-212-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR