Dinonylphthalat

Lieferant: EHRENSTORFER

C16174800
EHERC16174800EA 46 EUR
EHERC16174800
Dinonylphthalat
Dinonylphthalat
Formel: C₂₆H₄₂O₄
MDL: MFCD00036237
CAS-Nummer: 84-76-4
EINECS: 201-560-0
UN: 3082
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR