Di-n-octylphthalat

Lieferant: EHRENSTORFER

C16175000
EHERC16175000EA 42 EUR
EHERC16175000
Di-n-octylphthalat
Di-n-octylphthalat
Formel: C₂₄H₃₈O₄
Molecular Weight: 390,56 g/mol
MDL: MFCD00015292
CAS-Nummer: 117-84-0
EINECS: 204-214-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR