Monomethylphthalat

Lieferant: EHRENSTORFER

Synonyme: 2-(Methoxycarbonyl)benzoic acid, Methyl hydrogen phthalate

C16179000
EHERC16179000EA 46 EUR
EHERC16179000
Monomethylphthalat
Monomethylphthalat
Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
MDL: MFCD00002466
CAS-Nummer: 4376-18-5
EINECS: 224-476-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR