Kupfer(II)phthalocyanin

Lieferant: EHRENSTORFER

Synonyme: [29H,31H-Phthalocyaninato-N29,N30,N31,N32]Kupfer, CuPc, Phthalocyanin-Kupfer(II)-Komplex, Phthalocyaninblau, Tetrabenzo-5,10,15,20-diazaporphyrinphthalocyanin, Pigment Blue 15, Kupfer-Phthalocyanin, Heliogenblau B

C16206500
EHERC16206500EA 97 EUR
EHERC16206500
Kupfer(II)phthalocyanin
Copper(II) phthalocyanine
Formel: C₃₂H₁₆CuN₈
Molecular Weight: 576,08 g/mol
Schmelzpunkt: 480 °C
MDL: MFCD00010719
CAS-Nummer: 147-14-8
EINECS: 205-685-1
Merck Index: 12,02584

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR