Procain Hydrochlorid

Lieferant: EHRENSTORFER

Synonyme: 2-Diethylaminoethyl 4-aminobenzoate hydrochloride

C16289500
EHERC16289500EA 76 EUR
EHERC16289500
Procain Hydrochlorid
Procain Hydrochlorid
Formel: C₁₃H₂₁ClN₂O₂
MDL: MFCD00013000
CAS-Nummer: 51-05-8
EINECS: 200-077-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR