Propham (Isopropyl-N-phenylcarbamat)

Lieferant: EHRENSTORFER

C16470000
EHERC16470000EA 61 EUR
EHERC16470000
Propham (Isopropyl-N-phenylcarbamat)
Propham (Isopropyl-N-phenylcarbamat)
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,22 g/mol
MDL: MFCD00026382
CAS-Nummer: 122-42-9
EINECS: 204-542-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR