Pyridoxin Hydrochlorid

Lieferant: EHRENSTORFER

Synonyme: Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI

C16652000
EHERC16652000EA 57 EUR
EHERC16652000
Pyridoxin Hydrochlorid
Pyridoxin Hydrochlorid
Formel: C₈H₁₂ClNO₃
Molecular Weight: 205,64 g/mol
Schmelzpunkt: 214…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012807
CAS-Nummer: 58-56-0
EINECS: 200-386-2
Merck Index: 13,08072

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR