2-Chinoxalincarbonsäure
Lieferant: EHRENSTORFER
C16713000
EHERC16713000EA
158
EUR
EHERC16713000
2-Chinoxalincarbonsäure
2-Quinoxalinecarboxylic acid
Formel:
C₉H₆N₂O₂ Molecular Weight: 174,16 g/mol |
MDL:
MFCD00012334 CAS-Nummer: 879-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...