D(+)-Raffinose Pentahydrat
Lieferant: EHRENSTORFER
Synonyme:
Melitose Pentahydrat, Melitriose Pentahydrat, Gossypose Pentahydrat
C16806000
EHERC16806000EA
40
EUR
EHERC16806000
D(+)-Raffinose Pentahydrat
D(+)-Raffinose Pentahydrat
Formel:
C₁₈H₃₂O₁₆·5H₂O Molecular Weight: 594.52 g/mol Schmelzpunkt: 80 Dichte: 1.516 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071590 CAS-Nummer: 17629-30-0 EINECS: 208-146-9 Merck Index: 12,08279 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...