Riboflavin (Vitamin B2)

Lieferant: EHRENSTORFER

C16813600
EHERC16813600EA 42 EUR
EHERC16813600
Riboflavin (Vitamin B2)
Riboflavin (Vitamin B2)
Formel: C₁₇H₂₀N₄O₆
Schmelzpunkt: 280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005022
CAS-Nummer: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR