Riboflavin-5'-phosphat Natriumsalz
Lieferant: EHRENSTORFER
Synonyme:
Vitamin B2 phosphate sodium salt, Riboflavin 5'-Monophosphate sodium salt, Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogen phosphate, Flavin mononucleotide, Lactoflavin phosphate sodium salt, Riboflavin phosphate sodium
C16813610
EHERC16813610EA
49
EUR
EHERC16813610
Riboflavin-5'-phosphat Natriumsalz
Riboflavin-5'-phosphat Natriumsalz
Formel:
C₁₇H₂₀N₄NaO₉P Molecular Weight: 478.3256 g/mol |
MDL:
MFCD00065362 CAS-Nummer: 130-40-5 EINECS: 204-988-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...