Saccharin
Lieferant: EHRENSTORFER
C16901000
EHERC16901000EA
28.49
EUR
EHERC16901000
Saccharin
Saccharin
Formel:
C₇H₅NO₃S |
MDL:
MFCD00005866 CAS-Nummer: 81-07-2 EINECS: 201-321-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...