Scopoletin (7-Hydroxy-6-methoxy-2H-chromen-2-on)

Lieferant: EHRENSTORFER

Synonyme: Scopoletin, 7-Hydroxy-6-methoxy-2H-chromen-2-one

C16916000
EHERC16916000EA 235 EUR
EHERC16916000
Scopoletin (7-Hydroxy-6-methoxy-2H-chromen-2-on)
Scopoletin (7-Hydroxy-6-methoxycumarin)
Formel: C₁₀H₈O₄
Molecular Weight: 192,1681 g/mol
MDL: MFCD00006872
CAS-Nummer: 92-61-5
EINECS: 202-171-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR