Simazin (2-Chlor-4,6-bis(ethylamino)-1,3,5-triazin)

Lieferant: EHRENSTORFER

C16950000
EHERC16950000EA 70 EUR
EHERC16950000
Simazin (2-Chlor-4,6-bis(ethylamino)-1,3,5-triazin)
Simazin (2-Chlor-4,6-bis(ethylamino)-1,3,5-triazin)
Formel: C₇H₁₂ClN₅
Molecular Weight: 201,66 g/mol
MDL: MFCD00023174
CAS-Nummer: 122-34-9
EINECS: 204-535-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR