1,4-Bis(p-toluidino)-9,10-anthrachinon
Lieferant: EHRENSTORFER
C16971100
EHERC16971100EA
100
EUR
EHERC16971100
1,4-Bis(p-toluidino)-9,10-anthrachinon
1,4-Bis(p-toluidino)-9,10-anthrachinon
Formel:
C₂₈H₂₂N₂O₂ |
MDL:
MFCD00001195 CAS-Nummer: 128-80-3 EINECS: 204-909-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...