1-Hydroxy-4-(p-toluidino)-9,10-anthrachinon
Lieferant: EHRENSTORFER
Synonyme:
1-Hydroxy-4-(p-tolylamino)anthracene-9,10-dione
C16971330
EHERC16971330EA
103
EUR
EHERC16971330
1-Hydroxy-4-(p-toluidino)-9,10-anthrachinon
1-Hydroxy-4-(p-toluidino)-9,10-anthrachinon
Formel:
C₂₁H₁₅NO₃ Molecular Weight: 329,3500 g/mol |
MDL:
MFCD00019148 CAS-Nummer: 81-48-1 EINECS: 201-353-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...