Spermin Tetrahydrochlorid
Lieferant: EHRENSTORFER
Synonyme:
N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine) tetrahydrochloride
C16972742
EHERC16972742EA
85
EUR
EHERC16972742
Spermin Tetrahydrochlorid
Spermin Tetrahydrochlorid
Formel:
C₁₀H₃₀Cl₄N₄ Molecular Weight: 348.19 g/mol |
MDL:
MFCD00012914 CAS-Nummer: 306-67-2 EINECS: 206-189-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...